Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qser1
Entrez Gene ID
311266
Gene Name
glutamine and serine rich 1
Gene Aliases
RGD1311804
Chromosome Location
Chr.3: 94702635 - 94767370 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139493.1 NP_001132965.1 2 - 3 208 96
XM_006234646.2 XP_006234708.1 2 - 3 311 96

View Full Product Details