Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc57
Entrez Gene ID
311346
Gene Name
leucine rich repeat containing 57
Gene Aliases
RGD1307128
Chromosome Location
Chr.3: 112365936 - 112371787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012354.2 NP_001012354.1 - 811 64
XM_006234777.2 XP_006234839.1 - 577 64
XM_006234778.1 XP_006234840.1 - 592 64
XM_006234779.2 XP_006234841.1 - 607 64
XM_006234780.2 XP_006234842.1 - 628 64
GenBank mRNA BC089966.1 - - 628 64

View Full Product Details