Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnrnpa3
Entrez Gene ID
362152
Gene Name
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases
Hnrpa3
Chromosome Location
Chr.3: 62481228 - 62491356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111294.1 NP_001104764.1 3 - 4 416 74
NM_001111295.1 NP_001104765.1 3 - 4 350 74
NM_198132.3 NP_937765.1 3 - 4 416 74
XM_008761911.1 XP_008760133.1 3 - 4 365 74
XM_008761912.1 XP_008760134.1 3 - 4 365 74
XM_008761913.1 XP_008760135.1 3 - 4 411 74
GenBank mRNA BC081878.1 - 3 - 4 356 74
FQ214617.1 - 3 - 4 312 74

View Full Product Details