Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tp53i11
Entrez Gene ID
311209
Gene Name
tumor protein p53 inducible protein 11
Gene Aliases
Trp53i11
Chromosome Location
Chr.3: 82081855 - 82097061 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107749.1 NP_001101219.1 4 - 5 430 83
XM_006234569.2 XP_006234631.1 4 - 5 658 83
XM_006234570.2 XP_006234632.1 4 - 5 619 83
XM_006234571.2 XP_006234633.1 3 - 4 285 83
XM_006234572.2 XP_006234634.1 4 - 5 468 83
GenBank mRNA BC167758.1 - 4 - 5 568 83

View Full Product Details