Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h15
Entrez Gene ID
362154
Gene Name
zinc finger CCCH-type containing 15
Gene Aliases
-
Chromosome Location
Chr.3: 71020426 - 71040159 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010963.1 NP_001010963.1 4 - 5 493 139
GenBank mRNA AY377983.1 - 4 - 5 449 139
BC088438.1 - 4 - 5 493 139
FQ232115.1 - 4 - 5 572 139
FQ233565.1 - 4 - 5 595 139

View Full Product Details