Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif3i
Entrez Gene ID
682390
Gene Name
eukaryotic translation initiation factor 3, subunit I
Gene Aliases
Eif3s2
Chromosome Location
Chr.5: 147796580 - 147803853 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115035.1 NP_001108507.1 - 290 82
GenBank mRNA BC158748.1 - - 258 82
BC158821.1 - - 286 82
FQ209543.1 - - 272 82
FQ220584.1 - - 274 82
FQ220807.1 - - 275 82
FQ227121.1 - - 276 82
FQ230070.1 - - 276 82
FQ231084.1 - - 266 82
FQ231314.1 - - 266 82

View Full Product Details