Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stk40
Entrez Gene ID
360230
Gene Name
serine/threonine kinase 40
Gene Aliases
Lyk4, RGD1307310
Chromosome Location
Chr.5: 144111931 - 144150653 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183056.2 NP_898879.2 8 - 9 1048 74
XM_006238881.2 XP_006238943.1 8 - 9 2758 74
GenBank mRNA AY336055.1 - 8 - 9 1050 74
BC100062.1 - 8 - 9 1048 74

View Full Product Details