Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk11b
Entrez Gene ID
252879
Gene Name
cyclin-dependent kinase 11B
Gene Aliases
Cdc2l1, Cdk11
Chromosome Location
Chr.5: 173051898 - 173078049 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145766.2 NP_665709.2 9 - 10 1059 117
XM_006239527.2 XP_006239589.1 8 - 9 1615 117
XM_006239528.2 XP_006239590.1 9 - 10 1422 117
XM_006239529.2 XP_006239591.1 9 - 10 1343 117
XM_006239530.1 XP_006239592.1 9 - 10 1043 117
XM_006239531.2 XP_006239593.1 9 - 10 1147 117
XM_006239532.2 XP_006239594.1 4 - 5 1491 117
XM_006239533.1 XP_006239595.1 4 - 5 468 117
GenBank mRNA DY556124.1 - 4 - 5 479 117
EX490221.1 - 1 - 2 149 117

View Full Product Details