Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smap2
Entrez Gene ID
298500
Gene Name
small ArfGAP2
Gene Aliases
RGD1308418, Smap1l
Chromosome Location
Chr.5: 139885321 - 139931366 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100669.1 NP_001094139.1 1 - 2 479 86
GenBank mRNA BC161927.1 - 1 - 2 473 86

View Full Product Details