Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcc2
Entrez Gene ID
298594
Gene Name
regulator of chromosome condensation 2
Gene Aliases
RGD1309986
Chromosome Location
Chr.5: 159220531 - 159236313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764324.1 XP_008762546.1 9 - 10 1047 71

View Full Product Details