Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oscp1
Entrez Gene ID
362595
Gene Name
organic solute carrier partner 1
Gene Aliases
RGD1306596
Chromosome Location
Chr.5: 144067466 - 144098073 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029923.1 NP_001025094.1 1 - 2 168 73
GenBank mRNA AB299031.1 - 1 - 2 71 73
BC098048.1 - 1 - 2 168 73

View Full Product Details