Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl20
Entrez Gene ID
680747
Gene Name
mitochondrial ribosomal protein L20
Gene Aliases
-
Chromosome Location
Chr.5: 173248245 - 173252775 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109428.1 NP_001102898.1 3 - 4 408 80
GenBank mRNA BC166541.1 - 3 - 4 293 80
FQ224518.1 - 3 - 4 312 80

View Full Product Details