The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031820.1 | NP_114008.1 | 6 - 7 | 706 | 52 |
XM_006239598.2 | XP_006239660.1 | 6 - 7 | 1020 | 52 | |
XM_008764388.1 | XP_008762610.1 | 6 - 7 | 1021 | 52 | |
XM_008764389.1 | XP_008762611.1 | 6 - 7 | 1021 | 52 | |
XM_008764390.1 | XP_008762612.1 | 6 - 7 | 1021 | 52 | |
XM_008764391.1 | XP_008762613.1 | 7 - 8 | 722 | 52 | |
GenBank mRNA | AF143545.2 | - | 6 - 7 | 706 | 52 |
AF143546.2 | - | 6 - 7 | 706 | 52 |