Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpf3
Entrez Gene ID
299186
Gene Name
D4, zinc and double PHD fingers, family 3
Gene Aliases
-
Chromosome Location
Chr.6: 106660569 - 106971299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191818.1 NP_001178747.1 2 - 3 221 109
XM_008764746.1 XP_008762968.1 3 - 4 441 109
XM_008764747.1 XP_008762969.1 4 - 5 422 109
XM_008764748.1 XP_008762970.1 3 - 4 440 109
XM_008764749.1 XP_008762971.1 2 - 3 270 109
XM_008764750.1 XP_008762972.1 3 - 4 440 109
XM_008764751.1 XP_008762973.1 2 - 3 687 109

View Full Product Details