Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd7
Entrez Gene ID
362772
Gene Name
BTB (POZ) domain containing 7
Gene Aliases
-
Chromosome Location
Chr.6: 126658398 - 126747883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108720.1 NP_001102190.1 5 - 6 1717 138
XM_008764818.1 XP_008763040.1 5 - 6 1752 138
XM_008764819.1 XP_008763041.1 6 - 7 1894 138

View Full Product Details