Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a9
Entrez Gene ID
314275
Gene Name
solute carrier family 39, member 9
Gene Aliases
-
Chromosome Location
Chr.6: 104290767 - 104326553 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034929.1 NP_001030101.1 4 - 5 887 71
XM_006240283.2 XP_006240345.1 4 - 5 1191 71
XM_008764784.1 XP_008763006.1 3 - 4 1320 71
GenBank mRNA BC105849.1 - 4 - 5 887 71

View Full Product Details