The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002835.1 | NP_001002835.1 | 11 - 12 | 1288 | 71 |
XM_006240284.2 | XP_006240346.1 | 11 - 12 | 1594 | 71 | |
XM_008764785.1 | XP_008763007.1 | 12 - 13 | 1694 | 71 | |
XM_008764786.1 | XP_008763008.1 | 12 - 13 | 1661 | 71 | |
GenBank mRNA | FQ222878.1 | - | 11 - 12 | 1545 | 71 |