Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smek1
Entrez Gene ID
314388
Gene Name
SMEK homolog 1, suppressor of mek1 (Dictyostelium)
Gene Aliases
RGD1309059
Chromosome Location
Chr.6: 125040803 - 125083496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108050.2 NP_001101520.2 11 - 12 2251 168
NM_001271174.1 NP_001258103.1 11 - 12 2548 168
GenBank mRNA DY321166.1 - 2 - 3 288 168

View Full Product Details