Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galc
Entrez Gene ID
314360
Gene Name
galactosylceramidase
Gene Aliases
-
Chromosome Location
Chr.6: 122177195 - 122239411 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005888.1 NP_001005888.1 11 - 12 1245 78
XM_008764799.1 XP_008763021.1 11 - 12 1681 78
GenBank mRNA AY386370.1 - 11 - 12 1245 78
FQ223346.1 - 11 - 12 1264 78
FQ228993.1 - 11 - 12 1264 78

View Full Product Details