Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Med6
Entrez Gene ID
299180
Gene Name
mediator complex subunit 6
Gene Aliases
-
Chromosome Location
Chr.6: 105302558 - 105316024 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106742.1 NP_001100212.1 5 - 6 535 138
GenBank mRNA BC158786.1 - 5 - 6 465 138

View Full Product Details