Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sfpq
Entrez Gene ID
252855
Gene Name
splicing factor proline/glutamine-rich
Gene Aliases
-
Chromosome Location
Chr.5: 145079803 - 145088517 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025271.1 NP_001020442.1 7 - 8 1862 68
GenBank mRNA BC098731.1 - 7 - 8 1862 68
FQ227268.1 - 3 - 4 327 68
FQ229853.1 - 3 - 4 404 68

View Full Product Details