Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pink1
Entrez Gene ID
298575
Gene Name
PTEN induced putative kinase 1
Gene Aliases
-
Chromosome Location
Chr.5: 156677146 - 156689258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106694.1 NP_001100164.1 3 - 4 754 65

View Full Product Details