Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb48
Entrez Gene ID
362668
Gene Name
zinc finger and BTB domain containing 48
Gene Aliases
Hkr3
Chromosome Location
Chr.5: 169191771 - 169200142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013216.2 NP_001013234.2 5 - 6 1226 83
XM_006239469.2 XP_006239531.1 5 - 6 1235 83
XM_008764367.1 XP_008762589.1 5 - 6 1264 83
GenBank mRNA BC091214.1 - 4 - 5 1825 83
FQ211844.1 - 5 - 6 1253 83

View Full Product Details