The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012746.1 | NP_001012764.1 | 2 - 3 | 431 | 83 |
XM_006239218.2 | XP_006239280.1 | 3 - 4 | 650 | 83 | |
GenBank mRNA | AY919305.1 | - | 2 - 3 | 431 | 83 |