Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wrap73
Entrez Gene ID
366515
Gene Name
WD repeat containing, antisense to TP73
Gene Aliases
Wdr8
Chromosome Location
Chr.5: 171441536 - 171455795 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014262.1 NP_001014284.1 8 - 9 834 72
GenBank mRNA BC082062.1 - 8 - 9 834 72

View Full Product Details