Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lsm10
Entrez Gene ID
366468
Gene Name
LSM10, U7 small nuclear RNA associated
Gene Aliases
-
Chromosome Location
Chr.5: 144106556 - 144108703 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238896.2 XP_006238958.1 1 - 2 926 79

View Full Product Details