Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc28b
Entrez Gene ID
682445
Gene Name
coiled coil domain containing 28B
Gene Aliases
-
Chromosome Location
Chr.5: 147823446 - 147830430 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134689.1 NP_001128161.1 3 - 4 404 80
XM_008764171.1 XP_008762393.1 3 - 4 3199 80
GenBank mRNA BC168155.1 - 2 - 3 781 80

View Full Product Details