Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhl3
Entrez Gene ID
313582
Gene Name
four and a half LIM domains 3
Gene Aliases
-
Chromosome Location
Chr.5: 142688970 - 142696330 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107979.1 NP_001101449.1 3 - 4 401 87
XM_008764016.1 XP_008762238.1 3 - 4 535 87
XM_008764017.1 XP_008762239.1 3 - 4 462 87

View Full Product Details