Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr61
Entrez Gene ID
310780
Gene Name
G protein-coupled receptor 61
Gene Aliases
-
Chromosome Location
Chr.2: 210937289 - 210943673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107715.2 NP_001101185.2 1 - 2 65 109

View Full Product Details