Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam73a
Entrez Gene ID
362058
Gene Name
family with sequence similarity 73, member A
Gene Aliases
RGD1559958
Chromosome Location
Chr.2: 257490729 - 257546850 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761559.1 XP_008759781.1 2 - 3 329 60

View Full Product Details