Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx7
Entrez Gene ID
310815
Gene Name
sorting nexin 7
Gene Aliases
-
Chromosome Location
Chr.2: 220008463 - 220027686 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012083.1 NP_001012083.1 1 - 2 165 153
XM_006233234.2 XP_006233296.1 1 - 2 1760 153
XM_006233235.2 XP_006233297.1 1 - 2 1761 153
GenBank mRNA BC082033.1 - 1 - 2 165 153

View Full Product Details