Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints12
Entrez Gene ID
295448
Gene Name
integrator complex subunit 12
Gene Aliases
Phf22
Chromosome Location
Chr.2: 238253119 - 238270722 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007640.1 NP_001007641.1 1 - 2 162 101
XM_006233320.2 XP_006233382.1 2 - 3 1292 101
XM_006233321.2 XP_006233383.1 2 - 3 278 101
GenBank mRNA BC079404.1 - 1 - 2 162 101
FQ212179.1 - 2 - 3 253 101

View Full Product Details