Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clca4
Entrez Gene ID
362053
Gene Name
chloride channel accessory 4
Gene Aliases
Clca6, Prp3
Chromosome Location
Chr.2: 250843165 - 250864060 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201419.1 NP_958822.1 6 - 7 857 80
GenBank mRNA AY333961.1 - 6 - 7 857 80

View Full Product Details