The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020091.1 | NP_064476.1 | 8 - 9 | 1022 | 66 |
GenBank mRNA | AJ251688.1 | - | 8 - 9 | 1022 | 66 |