The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106476.1 | NP_001099946.1 | 6 - 7 | 605 | 104 |
GenBank mRNA | FQ225038.1 | - | 6 - 7 | 528 | 104 |
FQ227458.1 | - | 6 - 7 | 528 | 104 |