Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dennd2d
Entrez Gene ID
310772
Gene Name
DENN/MADD domain containing 2D
Gene Aliases
RGD1305120
Chromosome Location
Chr.2: 209097894 - 209116971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107714.1 NP_001101184.1 4 - 5 502 92
XM_006233114.2 XP_006233176.1 4 - 5 451 92
XM_008761396.1 XP_008759618.1 3 - 4 898 92
XM_008761397.1 XP_008759619.1 3 - 4 898 92
XM_008761398.1 XP_008759620.1 4 - 5 451 92
XM_008761399.1 XP_008759621.1 4 - 5 535 92
GenBank mRNA BC158880.1 - 4 - 5 438 92

View Full Product Details