Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kat2b
Entrez Gene ID
301164
Gene Name
K(lysine) acetyltransferase 2B
Gene Aliases
Pcaf
Chromosome Location
Chr.9: 4440982 - 4493791 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750617.3 XP_003750665.2 5 - 6 1861 88

View Full Product Details