The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030858.1 | NP_110485.1 | 3 - 4 | 2168 | 132 |
XM_006254959.2 | XP_006255021.1 | 3 - 4 | 762 | 132 | |
GenBank mRNA | AF042499.1 | - | 3 - 4 | 890 | 132 |
AF159626.1 | - | 3 - 4 | 2168 | 132 |