Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sult1d1
Entrez Gene ID
60393
Gene Name
sulfotransferase family 1D, member 1
Gene Aliases
ST1D1, Sult-n, Sultn
Chromosome Location
Chr.14: 22107418 - 22126011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021769.1 NP_068537.1 3 - 4 97
GenBank mRNA U32372.1 - 3 - 4 97

View Full Product Details