Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx19
Entrez Gene ID
315478
Gene Name
sorting nexin 19
Gene Aliases
-
Chromosome Location
Chr.8: 31497031 - 31534077 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108131.1 NP_001101601.1 1 - 2 1548 108
XM_006242747.2 XP_006242809.1 2 - 3 1865 108

View Full Product Details