Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eepd1
Entrez Gene ID
315500
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
RGD1307401
Chromosome Location
Chr.8: 26651264 - 26758330 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014088.3 NP_001014110.1 3 - 4 1095 130
XM_006242726.2 XP_006242788.1 4 - 5 1529 130
GenBank mRNA BC083816.1 - 3 - 4 1095 130
BC099214.1 - 4 - 5 1487 130

View Full Product Details