Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb44
Entrez Gene ID
363035
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
Btbd15, RGD1308984
Chromosome Location
Chr.8: 32165623 - 32223341 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034942.1 NP_001030114.1 3 - 4 1266 130
XM_006242768.2 XP_006242830.1 3 - 4 1453 130
GenBank mRNA BC104680.1 - 3 - 4 1266 130

View Full Product Details