Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epdr1
Entrez Gene ID
291180
Gene Name
ependymin related 1
Gene Aliases
Epdr2, MERP2
Chromosome Location
Chr.17: 47397558 - 47422183 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007625.1 NP_001007626.1 2 - 3 692 71
GenBank mRNA BC083701.1 - 2 - 3 692 71

View Full Product Details