Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddah2
Entrez Gene ID
294239
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
-
Chromosome Location
Chr.20: 5049265 - 5052573 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772715.1 XP_008770937.1 5 - 6 1101 74

View Full Product Details