The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031318.1 | NP_112608.1 | 2 - 3 | 85 | 73 |
XM_006227901.2 | XP_006227963.1 | 2 - 3 | 150 | 73 | |
GenBank mRNA | AB010119.1 | - | 2 - 3 | 74 | 73 |
AJ131437.1 | - | 2 - 3 | 85 | 73 | |
BC166879.1 | - | 2 - 3 | 117 | 73 |