The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751731.3 | XP_003751779.1 | - | 1914 | 82 |
XM_008771919.1 | XP_008770141.1 | - | 2094 | 82 | |
XM_008771920.1 | XP_008770142.1 | - | 2088 | 82 | |
XM_008771921.1 | XP_008770143.1 | - | 1920 | 82 | |
GenBank mRNA | BC079322.1 | - | - | 1080 | 82 |