Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Setbp1
Entrez Gene ID
291423
Gene Name
SET binding protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 75095408 - 75267307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772236.1 XP_008770458.1 2 - 3 7435 119

View Full Product Details