Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc39a5
Entrez Gene ID
362812
Gene Name
solute carrier family 39 (zinc transporter), member 5
Gene Aliases
-
Chromosome Location
Chr.7: 2813875 - 2819906 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108728.1 NP_001102198.1 8 - 9 1560 109
XM_006240760.2 XP_006240822.1 9 - 10 1465 109
XM_006240761.2 XP_006240823.1 9 - 10 1477 109
XM_006240762.2 XP_006240824.1 10 - 11 1585 109
XM_006240763.2 XP_006240825.1 10 - 11 1573 109
XM_006240764.2 XP_006240826.1 8 - 9 1369 109
XM_006240766.2 XP_006240828.1 9 - 10 1680 109
XM_006240767.2 XP_006240829.1 7 - 8 1129 109
XM_008765027.1 XP_008763249.1 9 - 10 1499 109
XM_008765028.1 XP_008763250.1 9 - 10 1465 109
GenBank mRNA FQ220010.1 - 5 - 6 655 109

View Full Product Details