Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbp1
Entrez Gene ID
24362
Gene Name
fructose-1,6-bisphosphatase 1
Gene Aliases
Fdp
Chromosome Location
Chr.17: 396175 - 417480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012558.3 NP_036690.2 4 - 5 783 66
GenBank mRNA BC078894.1 - 4 - 5 765 66
BC078895.1 - 4 - 5 783 66
J04112.1 - 4 - 5 579 66
M86240.1 - 4 - 5 669 66

View Full Product Details