Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Habp4
Entrez Gene ID
361196
Gene Name
hyaluronan binding protein 4
Gene Aliases
-
Chromosome Location
Chr.17: 1679562 - 1702251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079940.1 NP_001073409.1 4 - 5 821 122
XM_006253494.2 XP_006253556.1 3 - 4 717 122
GenBank mRNA BC129111.1 - 4 - 5 821 122

View Full Product Details